Gene Transcripts
- Gene Symbol
- HIST1H2BD
- Entrez Gene ID
- 3017
- Gene Name
- histone cluster 1, H2bd
- Gene Aliases
- H2B.1B, H2B/b, H2BFB, HIRIP2, dJ221C16.6
- Chromosome Location
- Chr. 6: 26157985 - 26171349 on Build GRCh38
- UniGene ID
- Hs.591797
- Species
- Homo sapiens
- Gene Symbol
- H2BFS
- Entrez Gene ID
- 54145
- Gene Name
- H2B histone family member S (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 21: 43565239 - 43565617 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC126339.1 |
- |
1 |
330 |
|
BC126369.1 |
- |
1 |
330 |
|
- Gene Symbol
- HIST1H2BC
- Entrez Gene ID
- 8347
- Gene Name
- histone cluster 1, H2bc
- Gene Aliases
- H2B.1, H2B/l, H2BFL, dJ221C16.3
- Chromosome Location
- Chr. 6: 26123467 - 26123904 on Build GRCh38
- UniGene ID
- Hs.658713
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BE
- Entrez Gene ID
- 8344
- Gene Name
- histone cluster 1, H2be
- Gene Aliases
- H2B.h, H2B/h, H2BFH, dJ221C16.8
- Chromosome Location
- Chr. 6: 26183796 - 26184230 on Build GRCh38
- UniGene ID
- Hs.534369
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BF
- Entrez Gene ID
- 8343
- Gene Name
- histone cluster 1, H2bf
- Gene Aliases
- H2B/g, H2BFG
- Chromosome Location
- Chr. 6: 26199559 - 26199988 on Build GRCh38
- UniGene ID
- Hs.182137
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BH
- Entrez Gene ID
- 8345
- Gene Name
- histone cluster 1, H2bh
- Gene Aliases
- H2B/j, H2BFJ
- Chromosome Location
- Chr. 6: 26251651 - 26252075 on Build GRCh38
- UniGene ID
- Hs.247815
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BJ
- Entrez Gene ID
- 8970
- Gene Name
- histone cluster 1, H2bj
- Gene Aliases
- H2B/r, H2BFR, H2BJ
- Chromosome Location
- Chr. 6: 27132316 - 27132796 on Build GRCh38
- UniGene ID
- Hs.591807
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BK
- Entrez Gene ID
- 85236
- Gene Name
- histone cluster 1, H2bk
- Gene Aliases
- H2B/S, H2BFAiii, H2BFT, H2BK
- Chromosome Location
- Chr. 6: 27138293 - 27146858 on Build GRCh38
- UniGene ID
- Hs.437275
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BL
- Entrez Gene ID
- 8340
- Gene Name
- histone cluster 1, H2bl
- Gene Aliases
- H2B/c, H2BFC, dJ97D16.4
- Chromosome Location
- Chr. 6: 27807479 - 27807931 on Build GRCh38
- UniGene ID
- Hs.137594
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BN
- Entrez Gene ID
- 8341
- Gene Name
- histone cluster 1, H2bn
- Gene Aliases
- H2B/d, H2BFD
- Chromosome Location
- Chr. 6: 27838662 - 27839110 on Build GRCh38
- UniGene ID
- Hs.534368
- Species
- Homo sapiens
- Gene Symbol
- HIST1H2BO
- Entrez Gene ID
- 8348
- Gene Name
- histone cluster 1, H2bo
- Gene Aliases
- H2B.2, H2B/n, H2BFN, dJ193B12.2
- Chromosome Location
- Chr. 6: 27893425 - 27893891 on Build GRCh38
- UniGene ID
- Hs.484991
- Species
- Homo sapiens
- Gene Symbol
- LOC102724334
- Entrez Gene ID
- 102724334
- Gene Name
- histone H2B type F-S-like
- Gene Aliases
- Chromosome Location
- Chr. 21: 5972923 - 5973357 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details