Gene Transcripts
- Gene Symbol
- FTL
- Entrez Gene ID
- 2512
- Gene Name
- ferritin light chain
- Gene Aliases
- LFTD, NBIA3
- Chromosome Location
- Chr. 19: 48965309 - 48966879 on Build GRCh38
- UniGene ID
- Hs.433670
- Species
- Homo sapiens
- Gene Symbol
- SLC4A1
- Entrez Gene ID
- 6521
- Gene Name
- solute carrier family 4 member 1 (Diego blood group)
- Gene Aliases
- AE1, BND3, CD233, CHC, DI, EMPB3, EPB3, FR, RTA1A, SAO, SPH4, SW, WD, WD1, WR
- Chromosome Location
- Chr. 17: 44248390 - 44268161 on Build GRCh38
- UniGene ID
- Hs.210751
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF113686.1 |
- |
|
1548 |
|
View Full Product Details