Gene Transcripts
- Gene Symbol
- C12orf50
- Entrez Gene ID
- 160419
- Gene Name
- chromosome 12 open reading frame 50
- Gene Aliases
- Chromosome Location
- Chr. 12: 87980039 - 88029399 on Build GRCh38
- UniGene ID
- Hs.112930
- Species
- Homo sapiens
- Gene Symbol
- ABCC9
- Entrez Gene ID
- 10060
- Gene Name
- ATP binding cassette subfamily C member 9
- Gene Aliases
- ABC37, ATFB12, CANTU, CMD1O, SUR2
- Chromosome Location
- Chr. 12: 21797389 - 21941869 on Build GRCh38
- UniGene ID
- Hs.732701
- Species
- Homo sapiens
- Gene Symbol
- CYSLTR1
- Entrez Gene ID
- 10800
- Gene Name
- cysteinyl leukotriene receptor 1
- Gene Aliases
- CYSLT1, CYSLT1R, CYSLTR, HMTMF81
- Chromosome Location
- Chr. X: 78271468 - 78327691 on Build GRCh38
- UniGene ID
- Hs.733809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023929.1 |
- |
2 |
2081 |
|
- Gene Symbol
- CYYR1-AS1
- Entrez Gene ID
- 100996571
- Gene Name
- cysteine and tyrosine rich 1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 21: 26393635 - 26569252 on Build GRCh38
- UniGene ID
- Hs.570432
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135515.1 |
|
4 |
3047 |
|
GenBank mRNA |
AK125338.1 |
- |
4 |
3047 |
|
JF693915.1 |
- |
4 |
2884 |
|
- Gene Symbol
- DDHD1
- Entrez Gene ID
- 80821
- Gene Name
- DDHD domain containing 1
- Gene Aliases
- PA-PLA1, PAPLA1, SPG28
- Chromosome Location
- Chr. 14: 53036740 - 53153328 on Build GRCh38
- UniGene ID
- Hs.125525
- Species
- Homo sapiens
- Gene Symbol
- DTNB
- Entrez Gene ID
- 1838
- Gene Name
- dystrobrevin beta
- Gene Aliases
- Chromosome Location
- Chr. 2: 25377220 - 25673647 on Build GRCh38
- UniGene ID
- Hs.307720
- Species
- Homo sapiens
- Gene Symbol
- ESR1
- Entrez Gene ID
- 2099
- Gene Name
- estrogen receptor 1
- Gene Aliases
- ER, ESR, ESRA, ESTRR, Era, NR3A1
- Chromosome Location
- Chr. 6: 151654148 - 152103274 on Build GRCh38
- UniGene ID
- Hs.208124
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M69297.1 |
- |
3 |
1584 |
|
- Gene Symbol
- FLJ22447
- Entrez Gene ID
- 400221
- Gene Name
- uncharacterized LOC400221
- Gene Aliases
- Chromosome Location
- Chr. 14: 61570540 - 61654713 on Build GRCh38
- UniGene ID
- Hs.344000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026100.1 |
- |
5 |
917 |
|
- Gene Symbol
- FNIP2
- Entrez Gene ID
- 57600
- Gene Name
- folliculin interacting protein 2
- Gene Aliases
- FNIPL, MAPO1
- Chromosome Location
- Chr. 4: 158768995 - 158908050 on Build GRCh38
- UniGene ID
- Hs.652441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC007861.1 |
- |
3 |
904 |
|
- Gene Symbol
- HELLPAR
- Entrez Gene ID
- 101101692
- Gene Name
- HELLP associated long non-coding RNA
- Gene Aliases
- LINC-HELLP, lncHELLP
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.160562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
JX088243.1 |
- |
1 |
43086 |
|
- Gene Symbol
- LCLAT1
- Entrez Gene ID
- 253558
- Gene Name
- lysocardiolipin acyltransferase 1
- Gene Aliases
- 1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849
- Chromosome Location
- Chr. 2: 30447226 - 30644225 on Build GRCh38
- UniGene ID
- Hs.468048
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095284.1 |
- |
7 |
1944 |
|
- Gene Symbol
- LINC00504
- Entrez Gene ID
- 201853
- Gene Name
- long intergenic non-protein coding RNA 504
- Gene Aliases
- Chromosome Location
- Chr. 4: 14470465 - 14888169 on Build GRCh38
- UniGene ID
- Hs.551051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126435.1 |
|
7 |
3208 |
|
- Gene Symbol
- LOC101927374
- Entrez Gene ID
- 101927374
- Gene Name
- uncharacterized LOC101927374
- Gene Aliases
- Chromosome Location
- Chr. 3: 79411727 - 79435227 on Build GRCh38
- UniGene ID
- Hs.570644
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110133.1 |
|
3 |
1521 |
|
GenBank mRNA |
AK094694.1 |
- |
3 |
1521 |
|
- Gene Symbol
- LOC101929567
- Entrez Gene ID
- 101929567
- Gene Name
- uncharacterized LOC101929567
- Gene Aliases
- Chromosome Location
- Chr. 2: 8559833 - 8583792 on Build GRCh38
- UniGene ID
- Hs.634706
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110257.1 |
|
6 |
6862 |
|
GenBank mRNA |
BX641130.1 |
- |
6 |
6869 |
|
- Gene Symbol
- LOC440716
- Entrez Gene ID
- 440716
- Gene Name
- uncharacterized LOC440716
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.640174
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126238.1 |
- |
1 |
1612 |
|
- Gene Symbol
- LOC643406
- Entrez Gene ID
- 643406
- Gene Name
- uncharacterized LOC643406
- Gene Aliases
- Chromosome Location
- Chr. 20: 5471196 - 5477134 on Build GRCh38
- UniGene ID
- Hs.431161
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029405.1 |
|
2 |
3083 |
|
GenBank mRNA |
AK125948.1 |
- |
2 |
3073 |
|
- Gene Symbol
- LVCAT5
- Entrez Gene ID
- 105375475
- Gene Name
- liver cancer-associated transcript 5
- Gene Aliases
- Chromosome Location
- Chr. 7: 119619430 - 119907375 on Build GRCh38
- UniGene ID
- Hs.566654
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_131963.1 |
|
3 |
824 |
|
- Gene Symbol
- RIC3
- Entrez Gene ID
- 79608
- Gene Name
- RIC3 acetylcholine receptor chaperone
- Gene Aliases
- AYST720, PRO1385
- Chromosome Location
- Chr. 11: 8092965 - 8169088 on Build GRCh38
- UniGene ID
- Hs.231850
- Species
- Homo sapiens
- Gene Symbol
- TRAPPC3L
- Entrez Gene ID
- 100128327
- Gene Name
- trafficking protein particle complex 3 like
- Gene Aliases
- BET3L, bA259P20.2
- Chromosome Location
- Chr. 6: 116496483 - 116545610 on Build GRCh38
- UniGene ID
- Hs.134795
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK002042.1 |
- |
3 |
1653 |
|
- Gene Symbol
- USP24
- Entrez Gene ID
- 23358
- Gene Name
- ubiquitin specific peptidase 24
- Gene Aliases
- Chromosome Location
- Chr. 1: 55066359 - 55215374 on Build GRCh38
- UniGene ID
- Hs.477009
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055871.1 |
- |
1 |
2589 |
|
- Gene Symbol
- WDPCP
- Entrez Gene ID
- 51057
- Gene Name
- WD repeat containing planar cell polarity effector
- Gene Aliases
- BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
- Chromosome Location
- Chr. 2: 63120755 - 63840825 on Build GRCh38
- UniGene ID
- Hs.414952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538331.1 |
- |
14 |
2725 |
|
View Full Product Details