Gene Transcripts
- Gene Symbol
- ATP9B
- Entrez Gene ID
- 374868
- Gene Name
- ATPase phospholipid transporting 9B (putative)
- Gene Aliases
- ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
- Chromosome Location
- Chr. 18: 79069275 - 79378391 on Build GRCh38
- UniGene ID
- Hs.465475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK295981.1 |
- |
5 |
793 |
|
BC053561.1 |
- |
5 |
711 |
|
- Gene Symbol
- CFAP54
- Entrez Gene ID
- 144535
- Gene Name
- cilia and flagella associated protein 54
- Gene Aliases
- C12orf55, C12orf63
- Chromosome Location
- Chr. 12: 96489566 - 96875555 on Build GRCh38
- UniGene ID
- Hs.382121
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041426.1 |
- |
7 |
1034 |
|
- Gene Symbol
- CYSLTR1
- Entrez Gene ID
- 10800
- Gene Name
- cysteinyl leukotriene receptor 1
- Gene Aliases
- CYSLT1, CYSLT1R, CYSLTR, HMTMF81
- Chromosome Location
- Chr. X: 78271468 - 78327691 on Build GRCh38
- UniGene ID
- Hs.733809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023929.1 |
- |
2 |
1110 |
|
- Gene Symbol
- EYS
- Entrez Gene ID
- 346007
- Gene Name
- eyes shut homolog (Drosophila)
- Gene Aliases
- C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
- Chromosome Location
- Chr. 6: 63719980 - 65707225 on Build GRCh38
- UniGene ID
- Hs.25067
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647741.1 |
- |
5 |
5099 |
|
- Gene Symbol
- KCNQ1OT1
- Entrez Gene ID
- 10984
- Gene Name
- KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
- Gene Aliases
- KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012
- Chromosome Location
- Chr. 11: 2608328 - 2699998 on Build GRCh38
- UniGene ID
- Hs.604823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002728.3 |
|
1 |
64108 |
|
- Gene Symbol
- LINC00504
- Entrez Gene ID
- 201853
- Gene Name
- long intergenic non-protein coding RNA 504
- Gene Aliases
- Chromosome Location
- Chr. 4: 14470465 - 14888169 on Build GRCh38
- UniGene ID
- Hs.551051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126435.1 |
|
7 |
2240 |
|
GenBank mRNA |
BC036758.2 |
- |
7 |
2252 |
|
- Gene Symbol
- RABGAP1L-IT1
- Entrez Gene ID
- 106480742
- Gene Name
- RABGAP1L intronic transcript 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.688745
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX485272.1 |
- |
2 |
553 |
|
- Gene Symbol
- RIC3
- Entrez Gene ID
- 79608
- Gene Name
- RIC3 acetylcholine receptor chaperone
- Gene Aliases
- AYST720, PRO1385
- Chromosome Location
- Chr. 11: 8092965 - 8169088 on Build GRCh38
- UniGene ID
- Hs.231850
- Species
- Homo sapiens
- Gene Symbol
- SMC5-AS1
- Entrez Gene ID
- 100507299
- Gene Name
- SMC5 antisense RNA 1 (head to head)
- Gene Aliases
- MAMDC2-AS2
- Chromosome Location
- Chr. 9: 70216059 - 70258874 on Build GRCh38
- UniGene ID
- Hs.556816
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR936772.1 |
- |
3 |
5898 |
|
- Gene Symbol
- TBC1D8B
- Entrez Gene ID
- 54885
- Gene Name
- TBC1 domain family member 8B
- Gene Aliases
- GRAMD8B
- Chromosome Location
- Chr. X: 106802689 - 106876150 on Build GRCh38
- UniGene ID
- Hs.351798
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648230.1 |
- |
7 |
3263 |
|
View Full Product Details