Gene Transcripts
- Gene Symbol
- ORC4
- Entrez Gene ID
- 5000
- Gene Name
- origin recognition complex subunit 4
- Gene Aliases
- ORC4L, ORC4P
- Chromosome Location
- Chr. 2: 147930397 - 148021604 on Build GRCh38
- UniGene ID
- Hs.558364
- Species
- Homo sapiens
- Gene Symbol
- ABCC9
- Entrez Gene ID
- 10060
- Gene Name
- ATP binding cassette subfamily C member 9
- Gene Aliases
- ABC37, ATFB12, CANTU, CMD1O, SUR2
- Chromosome Location
- Chr. 12: 21797389 - 21941869 on Build GRCh38
- UniGene ID
- Hs.732701
- Species
- Homo sapiens
- Gene Symbol
- ADGRE4P
- Entrez Gene ID
- 326342
- Gene Name
- adhesion G protein-coupled receptor E4, pseudogene
- Gene Aliases
- EMR4, EMR4P, FIRE, GPR127, PGR16
- Chromosome Location
- Chr. 19: 6952500 - 6990846 on Build GRCh38
- UniGene ID
- Hs.688043
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024075.1 |
|
16 |
2523 |
|
GenBank mRNA |
AF489700.1 |
- |
16 |
2523 |
|
- Gene Symbol
- C12orf50
- Entrez Gene ID
- 160419
- Gene Name
- chromosome 12 open reading frame 50
- Gene Aliases
- Chromosome Location
- Chr. 12: 87980039 - 88029399 on Build GRCh38
- UniGene ID
- Hs.112930
- Species
- Homo sapiens
- Gene Symbol
- DTNB
- Entrez Gene ID
- 1838
- Gene Name
- dystrobrevin beta
- Gene Aliases
- Chromosome Location
- Chr. 2: 25377220 - 25673647 on Build GRCh38
- UniGene ID
- Hs.307720
- Species
- Homo sapiens
- Gene Symbol
- ESR1
- Entrez Gene ID
- 2099
- Gene Name
- estrogen receptor 1
- Gene Aliases
- ER, ESR, ESRA, ESTRR, Era, NR3A1
- Chromosome Location
- Chr. 6: 151654148 - 152103274 on Build GRCh38
- UniGene ID
- Hs.208124
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M69297.1 |
- |
3 |
1668 |
|
- Gene Symbol
- FLJ22447
- Entrez Gene ID
- 400221
- Gene Name
- uncharacterized LOC400221
- Gene Aliases
- Chromosome Location
- Chr. 14: 61570540 - 61654713 on Build GRCh38
- UniGene ID
- Hs.344000
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026100.1 |
- |
5 |
1001 |
|
- Gene Symbol
- FNIP2
- Entrez Gene ID
- 57600
- Gene Name
- folliculin interacting protein 2
- Gene Aliases
- FNIPL, MAPO1
- Chromosome Location
- Chr. 4: 158768995 - 158908050 on Build GRCh38
- UniGene ID
- Hs.652441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC007861.1 |
- |
3 |
988 |
|
- Gene Symbol
- L2HGDH
- Entrez Gene ID
- 79944
- Gene Name
- L-2-hydroxyglutarate dehydrogenase
- Gene Aliases
- C14orf160, L2HGA
- Chromosome Location
- Chr. 14: 50242434 - 50312235 on Build GRCh38
- UniGene ID
- Hs.256034
- Species
- Homo sapiens
- Gene Symbol
- LINC00504
- Entrez Gene ID
- 201853
- Gene Name
- long intergenic non-protein coding RNA 504
- Gene Aliases
- Chromosome Location
- Chr. 4: 14470465 - 14888169 on Build GRCh38
- UniGene ID
- Hs.551051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126435.1 |
|
7 |
3296 |
|
- Gene Symbol
- LOC101929567
- Entrez Gene ID
- 101929567
- Gene Name
- uncharacterized LOC101929567
- Gene Aliases
- Chromosome Location
- Chr. 2: 8559833 - 8583792 on Build GRCh38
- UniGene ID
- Hs.634706
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110257.1 |
|
6 |
6945 |
|
GenBank mRNA |
BX641130.1 |
- |
6 |
6952 |
|
- Gene Symbol
- LOC285690
- Entrez Gene ID
- 285690
- Gene Name
- uncharacterized LOC285690
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.633797
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094742.1 |
- |
1 |
1796 |
|
- Gene Symbol
- LOC440716
- Entrez Gene ID
- 440716
- Gene Name
- uncharacterized LOC440716
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.640174
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126238.1 |
- |
1 |
1696 |
|
- Gene Symbol
- RIC3
- Entrez Gene ID
- 79608
- Gene Name
- RIC3 acetylcholine receptor chaperone
- Gene Aliases
- AYST720, PRO1385
- Chromosome Location
- Chr. 11: 8092965 - 8169088 on Build GRCh38
- UniGene ID
- Hs.231850
- Species
- Homo sapiens
- Gene Symbol
- SMC5-AS1
- Entrez Gene ID
- 100507299
- Gene Name
- SMC5 antisense RNA 1 (head to head)
- Gene Aliases
- MAMDC2-AS2
- Chromosome Location
- Chr. 9: 70216059 - 70258874 on Build GRCh38
- UniGene ID
- Hs.556816
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR936772.1 |
- |
3 |
6957 |
|
- Gene Symbol
- TCF12
- Entrez Gene ID
- 6938
- Gene Name
- transcription factor 12
- Gene Aliases
- CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
- Chromosome Location
- Chr. 15: 56918090 - 57291261 on Build GRCh38
- UniGene ID
- Hs.511504
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647333.1 |
- |
1 |
7053 |
|
- Gene Symbol
- USP24
- Entrez Gene ID
- 23358
- Gene Name
- ubiquitin specific peptidase 24
- Gene Aliases
- Chromosome Location
- Chr. 1: 55066359 - 55215374 on Build GRCh38
- UniGene ID
- Hs.477009
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055871.1 |
- |
1 |
2673 |
|
View Full Product Details