Gene Transcripts

Gene Symbol
RLIM
Entrez Gene ID
51132
Gene Name
ring finger protein, LIM domain interacting
Gene Aliases
NY-REN-43, RNF12
Chromosome Location
Chr. X: 74582976 - 74614626 on Build GRCh38
UniGene ID
Hs.653288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016120.3 NP_057204.2 4 10134
NM_183353.2 NP_899196.1 5 10196
GenBank mRNA AK127880.1 - 1 1983

View Full Product Details