Gene Transcripts

Gene Symbol
CCL3L3
Entrez Gene ID
414062
Gene Name
C-C motif chemokine ligand 3 like 3
Gene Aliases
464.2, D17S1718, G0S19-2, LD78, LD78BETA, SCYA3L, SCYA3L1
Chromosome Location
Chr. 17: 36194869 - 36196748 on Build GRCh38
UniGene ID
Hs.454713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001437.3 NP_001001437.2 1 28
GenBank mRNA AA380745.1 - 1 28
BC007783.2 - 1 23 4308472

Gene Symbol
CCL3L1
Entrez Gene ID
6349
Gene Name
C-C motif chemokine ligand 3 like 1
Gene Aliases
464.2, D17S1718, G0S19-2, LD78, LD78-beta(1-70), LD78BETA, MIP1AP, SCYA3L, SCYA3L1
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.512683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021006.5 NP_066286.1 1 38
NR_111964.1 1 38
GenBank mRNA AV739733.1 - 1 82
BC027888.1 - 1 26 5222247
BC107710.1 - 1 42 6699133
X52149.1 - 1 24

View Full Product Details