Gene Transcripts
- Gene Symbol
- LINC00504
- Entrez Gene ID
- 201853
- Gene Name
- long intergenic non-protein coding RNA 504
- Gene Aliases
- Chromosome Location
- Chr. 4: 14470465 - 14888169 on Build GRCh38
- UniGene ID
- Hs.551051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126435.1 |
|
7 |
2651 |
|
GenBank mRNA |
BC036758.2 |
- |
7 |
2663 |
|
- Gene Symbol
- ATP9B
- Entrez Gene ID
- 374868
- Gene Name
- ATPase phospholipid transporting 9B (putative)
- Gene Aliases
- ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
- Chromosome Location
- Chr. 18: 79069275 - 79378391 on Build GRCh38
- UniGene ID
- Hs.465475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC053561.1 |
- |
5 |
1122 |
|
- Gene Symbol
- CFAP54
- Entrez Gene ID
- 144535
- Gene Name
- cilia and flagella associated protein 54
- Gene Aliases
- C12orf55, C12orf63
- Chromosome Location
- Chr. 12: 96489566 - 96875555 on Build GRCh38
- UniGene ID
- Hs.382121
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041426.1 |
- |
7 |
1445 |
|
- Gene Symbol
- LOC101926898
- Entrez Gene ID
- 101926898
- Gene Name
- uncharacterized LOC101926898
- Gene Aliases
- Chromosome Location
- Chr. 6: 46492052 - 46533486 on Build GRCh38
- UniGene ID
- Hs.639416
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_125837.1 |
|
3 |
903 |
|
GenBank mRNA |
BC042616.1 |
- |
3 |
905 |
|
- Gene Symbol
- LOC101929163
- Entrez Gene ID
- 101929163
- Gene Name
- uncharacterized LOC101929163
- Gene Aliases
- Chromosome Location
- Chr. 6: 32255173 - 32407730 on Build GRCh38
- UniGene ID
- Hs.57692
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123889.1 |
- |
2 |
3232 |
|
- Gene Symbol
- RIC3
- Entrez Gene ID
- 79608
- Gene Name
- RIC3 acetylcholine receptor chaperone
- Gene Aliases
- AYST720, PRO1385
- Chromosome Location
- Chr. 11: 8092965 - 8169088 on Build GRCh38
- UniGene ID
- Hs.231850
- Species
- Homo sapiens
- Gene Symbol
- TBC1D8B
- Entrez Gene ID
- 54885
- Gene Name
- TBC1 domain family member 8B
- Gene Aliases
- GRAMD8B
- Chromosome Location
- Chr. X: 106802689 - 106876150 on Build GRCh38
- UniGene ID
- Hs.351798
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648230.1 |
- |
7 |
3674 |
|
- Gene Symbol
- WDPCP
- Entrez Gene ID
- 51057
- Gene Name
- WD repeat containing planar cell polarity effector
- Gene Aliases
- BBS15, C2orf86, CHDTHP, FRITZ, FRTZ
- Chromosome Location
- Chr. 2: 63120755 - 63840825 on Build GRCh38
- UniGene ID
- Hs.414952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX538331.1 |
- |
14 |
2162 |
|
View Full Product Details