Gene Transcripts

Gene Symbol
HMCN2
Entrez Gene ID
256158
Gene Name
hemicentin 2
Gene Aliases
Chromosome Location
Chr. 9: 130265742 - 130434123 on Build GRCh38
UniGene ID
Hs.32194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291815.1 NP_001278744.1 21 3175
XM_011518465.2 XP_011516767.1 21 3312
XM_011518466.2 XP_011516768.1 21 3312
XM_011518467.2 XP_011516769.1 21 3312
XM_011518468.2 XP_011516770.1 21 3312
XM_011518469.2 XP_011516771.1 21 3312
XM_011518470.2 XP_011516772.1 21 3312
GenBank mRNA AK128020.1 - 9 1135
BC146865.1 - 8 1023 9056793

View Full Product Details