Gene Transcripts

Gene Symbol
Tmem263
Entrez Gene ID
103266
Gene Name
transmembrane protein 263
Gene Aliases
AI597468
Chromosome Location
Chr. 10: 85102627 - 85117747 on Build GRCm38
UniGene ID
Mm.335025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013028.2 NP_001013046.1 3 3275
GenBank mRNA BC086492.1 - 3 3139

View Full Product Details