Gene Transcripts
- Gene Symbol
- ARMC4
- Entrez Gene ID
- 55130
- Gene Name
- armadillo repeat containing 4
- Gene Aliases
- CILD23
- Chromosome Location
- Chr. 10: 27796355 - 27999050 on Build GRCh38
- UniGene ID
- Hs.127530
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001238.1 |
- |
6 |
736 |
|
View Full Product Details