Gene Transcripts
- Gene Symbol
- Olfr738
- Entrez Gene ID
- 258662
- Gene Name
- olfactory receptor 738
- Gene Aliases
- MOR106-3
- Chromosome Location
- Chr. 14: 50413546 - 50414481 on Build GRCm38
- UniGene ID
- Mm.377625
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_146420.2 |
NP_666532.2
|
1 |
608 |
|
View Full Product Details