Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028718.2 | NP_082994.1 | 1, 2 | 225 | |
GenBank mRNA | AY613437.1 | - | 1, 2 | 228 | |
BC046538.1 | - | 1, 2 | 228 | ||
BC117868.2 | - | 1, 2 | 199 | 40095476 | |
BY739359.1 | - | 1, 2 | 225 |