Gene Transcripts

Gene Symbol
LOC728485
Entrez Gene ID
728485
Gene Name
uncharacterized LOC728485
Gene Aliases
Chromosome Location
Chr. 19: 36773118 - 36777078 on Build GRCh38
UniGene ID
Hs.157101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294306.1 NP_001281235.1 2 1563
GenBank mRNA AK092138.1 - 2 1542

View Full Product Details