Gene Transcripts

Gene Symbol
IFNK
Entrez Gene ID
56832
Gene Name
interferon kappa
Gene Aliases
IFNT1, INFE1
Chromosome Location
Chr. 9: 27524314 - 27526498 on Build GRCh38
UniGene ID
Hs.591083
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020124.2 NP_064509.2 1 273
GenBank mRNA AF146759.1 - 1 295
AF315688.1 - 1 284
AY358855.1 - 1 251

View Full Product Details