Gene Transcripts

Gene Symbol
Zfp943
Entrez Gene ID
74670
Gene Name
zinc finger prtoein 943
Gene Aliases
4930432O21Rik
Chromosome Location
Chr. 17: 21962559 - 21994366 on Build GRCm38
UniGene ID
Mm.26540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025373.2 NP_001020544.1 4 1312
GenBank mRNA BC066031.1 - 4 1113

View Full Product Details