Gene Transcripts

Gene Symbol
H60a
Entrez Gene ID
15101
Gene Name
histocompatibility 60a
Gene Aliases
H60
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.387042
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010400.2 NP_034530.1 3926
GenBank mRNA AK169909.1 - 3928

View Full Product Details