Gene Transcripts

Gene Symbol
IFNB1
Entrez Gene ID
3456
Gene Name
interferon beta 1
Gene Aliases
IFB, IFF, IFN-beta, IFNB
Chromosome Location
Chr. 9: 21077105 - 21077963 on Build GRCh38
UniGene ID
Hs.93177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB451323.1 - 1 151
AB451452.1 - 1 151
AB451491.1 - 1 88
BC096151.2 - 1 151 40002941
V00547.1 - 1 151

View Full Product Details