Gene Transcripts
- Gene Symbol
- RELA
- Entrez Gene ID
- 5970
- Gene Name
- RELA proto-oncogene, NF-kB subunit
- Gene Aliases
- NFKB3, p65
- Chromosome Location
- Chr. 11: 65653596 - 65662972 on Build GRCh38
- UniGene ID
- Hs.502875
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145138.1 |
NP_001138610.1
|
7, 8 |
793 |
|
NM_001243984.1 |
NP_001230913.1
|
7, 8 |
802 |
|
NM_001243985.1 |
NP_001230914.1
|
7, 8 |
802 |
|
NM_021975.3 |
NP_068810.3
|
7, 8 |
802 |
|
GenBank mRNA |
AK303262.1 |
- |
7, 8 |
781 |
|
AK308020.1 |
- |
7, 8 |
746 |
|
AK308478.1 |
- |
6, 7 |
848 |
|
BC014095.2 |
- |
7, 8 |
734 |
|
BC033522.1 |
- |
6, 7 |
659 |
5162908 |
BC110830.1 |
- |
7, 8 |
725 |
6019711 |
BM916512.1 |
- |
7, 8 |
723 |
|
L19067.1 |
- |
6, 7 |
661 |
|
M62399.1 |
- |
7, 8 |
741 |
|
View Full Product Details