Gene Transcripts
- Gene Symbol
- FAM153A
- Entrez Gene ID
- 285596
- Gene Name
- family with sequence similarity 153 member A
- Gene Aliases
- NY-REN-7
- Chromosome Location
- Chr. 5: 177694247 - 177783518 on Build GRCh38
- UniGene ID
- Hs.368516
- Species
- Homo sapiens
- Gene Symbol
- FAM153B
- Entrez Gene ID
- 202134
- Gene Name
- family with sequence similarity 153 member B
- Gene Aliases
- Chromosome Location
- Chr. 5: 176063709 - 176114798 on Build GRCh38
- UniGene ID
- Hs.368516
- Species
- Homo sapiens
View Full Product Details