Gene Transcripts

Gene Symbol
Ifna7
Entrez Gene ID
15970
Gene Name
interferon alpha 7
Gene Aliases
Ifa7
Chromosome Location
Chr. 4: 88816225 - 88816800 on Build GRCm38
UniGene ID
Mm.377091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008334.3 NP_032360.2 1 562
GenBank mRNA BC130231.1 - 1 563

View Full Product Details