Gene Transcripts

Gene Symbol
Uhrf1bp1
Entrez Gene ID
224648
Gene Name
UHRF1 (ICBP90) binding protein 1
Gene Aliases
1110020K19Rik, AA408674, F830021D11Rik, mKIAA4127
Chromosome Location
Chr. 17: 27856507 - 27900040 on Build GRCm38
UniGene ID
Mm.291291
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080769.1 NP_001074238.1 14 2708
GenBank mRNA AK089790.1 - 14 2735
AK144200.1 - 9 2004
AK220293.1 - 14 2708

View Full Product Details