Gene Transcripts

Gene Symbol
Jtb
Entrez Gene ID
29439
Gene Name
jumping translocation breakpoint
Gene Aliases
Chromosome Location
Chr. 2: 189591707 - 189595926 on Build Rnor_6.0
UniGene ID
Rn.91803
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019213.2 NP_062086.1 2, 3 388
GenBank mRNA AB016489.1 - 2, 3 442
BC062090.1 - 2, 3 388 5621612

View Full Product Details