Gene Transcripts

Gene Symbol
4930402F06Rik
Entrez Gene ID
74854
Gene Name
RIKEN cDNA 4930402F06 gene
Gene Aliases
Chromosome Location
Chr. 2: 35375562 - 35397174 on Build GRCm38
UniGene ID
Mm.159338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080709.2 NP_001074178.1 1, 2 277
GenBank mRNA AK015047.1 - 1, 2 278

View Full Product Details