Gene Transcripts

Gene Symbol
Nr1h2
Entrez Gene ID
58851
Gene Name
nuclear receptor subfamily 1, group H, member 2
Gene Aliases
OR-1
Chromosome Location
Chr. 1: 100554577 - 100559896 on Build Rnor_6.0
UniGene ID
Rn.786
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031626.1 NP_113814.1 6 713
GenBank mRNA BC157812.1 - 6 692
U14533.1 - 6 713
U20389.1 - 6 657

View Full Product Details