Gene Transcripts

Gene Symbol
TGFBR1
Entrez Gene ID
7046
Gene Name
transforming growth factor beta receptor 1
Gene Aliases
AAT5, ACVRLK4, ALK-5, ALK5, ESS1, LDS1, LDS1A, LDS2A, MSSE, SKR4, TGFR-1, tbetaR-I
Chromosome Location
Chr. 9: 99104038 - 99154192 on Build GRCh38
UniGene ID
Hs.494622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306210.1 NP_001293139.1 3 656
NM_004612.3 NP_004603.1 3 644
XM_011518948.2 XP_011517250.1 3 571
XM_011518949.2 XP_011517251.1 3 499
XM_017015063.1 XP_016870552.1 3 534
GenBank mRNA AJ619019.2 - 3 615
AK302059.1 - 3 644
AK302234.1 - 3 517
L11695.1 - 3 603

View Full Product Details