Gene Transcripts
- Gene Symbol
- Dlx4
- Entrez Gene ID
- 13394
- Gene Name
- distal-less homeobox 4
- Gene Aliases
- Dlx-4, Dlx7
- Chromosome Location
- Chr. 11: 95140447 - 95146499 on Build GRCm38
- UniGene ID
- Mm.439766
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007867.4 |
NP_031893.3
|
1, 2 |
601 |
|
GenBank mRNA |
U73329.1 |
- |
1, 2 |
281 |
|
View Full Product Details