Gene Transcripts
- Gene Symbol
- Polr2c
- Entrez Gene ID
- 20021
- Gene Name
- polymerase (RNA) II (DNA directed) polypeptide C
- Gene Aliases
- 33kDa, RPB3, Rpo2-3, mRBP31
- Chromosome Location
- Chr. 8: 94857450 - 94864242 on Build GRCm38
- UniGene ID
- Mm.292492
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC002023.1 |
- |
9 |
1013 |
|
View Full Product Details