Gene Transcripts
- Gene Symbol
- Oas1b
- Entrez Gene ID
- 23961
- Gene Name
- 2'-5' oligoadenylate synthetase 1B
- Gene Aliases
- Flv, L1, Mmu-L1, Oas1, Oias-2, Oias2, Wnv
- Chromosome Location
- Chr. 5: 120812635 - 120824163 on Build GRCm38
- UniGene ID
- Mm.233471
- Species
- Mus musculus
- Gene Symbol
- 1700007M16Rik
- Entrez Gene ID
- 72237
- Gene Name
- RIKEN cDNA 1700007M16 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK005735.1 |
- |
1 |
33 |
|
- Gene Symbol
- 2210020O09Rik
- Entrez Gene ID
- 72359
- Gene Name
- RIKEN cDNA 2210020O09 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008758.1 |
- |
1 |
23 |
|
- Gene Symbol
- 4833415N18Rik
- Entrez Gene ID
- 73761
- Gene Name
- RIKEN cDNA 4833415N18 gene
- Gene Aliases
- Chromosome Location
- Chr. 16: 11779702 - 11781501 on Build GRCm38
- UniGene ID
- Mm.39183
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BE653989.1 |
- |
1 |
59 |
|
- Gene Symbol
- 4921518K17Rik
- Entrez Gene ID
- 78758
- Gene Name
- RIKEN cDNA 4921518K17 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 57585984 - 57617511 on Build GRCm38
- UniGene ID
- Mm.160125
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019540.1 |
- |
|
1574 |
|
- Gene Symbol
- 4930421J07Rik
- Entrez Gene ID
- 73953
- Gene Name
- RIKEN cDNA 4930421J07 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.475762
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK149238.1 |
- |
1 |
1706 |
|
- Gene Symbol
- 5430427M07Rik
- Entrez Gene ID
- 71349
- Gene Name
- RIKEN cDNA 5430427M07 gene
- Gene Aliases
- Chromosome Location
- Chr. 12: 91045848 - 91056636 on Build GRCm38
- UniGene ID
- Mm.442093
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK017349.1 |
- |
1 |
33 |
|
- Gene Symbol
- 9130209A04Rik
- Entrez Gene ID
- 77668
- Gene Name
- RIKEN cDNA 9130209A04 gene
- Gene Aliases
- Chromosome Location
- Chr. 18: 47452668 - 47470868 on Build GRCm38
- UniGene ID
- Mm.98433
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020276.1 |
- |
1 |
32 |
|
- Gene Symbol
- 9130221F21Rik
- Entrez Gene ID
- 330350
- Gene Name
- RIKEN cDNA 9130221F21 gene
- Gene Aliases
- 9130221F21
- Chromosome Location
- Chr. 6: 72013896 - 72042528 on Build GRCm38
- UniGene ID
- Mm.246787
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK033688.1 |
- |
3 |
1353 |
|
- Gene Symbol
- Ano6
- Entrez Gene ID
- 105722
- Gene Name
- anoctamin 6
- Gene Aliases
- 2900059G15Rik, AA407480, AW554778, F730003B03Rik, Tmem16f
- Chromosome Location
- Chr. 15: 95790843 - 95975472 on Build GRCm38
- UniGene ID
- Mm.38087
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK144584.1 |
- |
2 |
2457 |
|
- Gene Symbol
- Auh
- Entrez Gene ID
- 11992
- Gene Name
- AU RNA binding protein/enoyl-coenzyme A hydratase
- Gene Aliases
- C77140, W91705
- Chromosome Location
- Chr. 13: 52835102 - 52929706 on Build GRCm38
- UniGene ID
- Mm.252034
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK169661.1 |
- |
4 |
1131 |
|
- Gene Symbol
- C130046K22Rik
- Entrez Gene ID
- 399609
- Gene Name
- RIKEN cDNA C130046K22 gene
- Gene Aliases
- Chromosome Location
- Chr. 11: 103697724 - 103725573 on Build GRCm38
- UniGene ID
- Mm.351922
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142001.1 |
- |
3 |
3333 |
|
- Gene Symbol
- C430002E04Rik
- Entrez Gene ID
- 78706
- Gene Name
- RIKEN cDNA C430002E04 gene
- Gene Aliases
- Chromosome Location
- Chr. 3: 41487681 - 41493199 on Build GRCm38
- UniGene ID
- Mm.366094
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK145605.1 |
- |
2 |
2040 |
|
- Gene Symbol
- Dynlt1f
- Entrez Gene ID
- 100040531
- Gene Name
- dynein light chain Tctex-type 1F
- Gene Aliases
- 100040631, Dynlt1e
- Chromosome Location
- Chr. 17: 6646606 - 6655939 on Build GRCm38
- UniGene ID
- Mm.457987
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK014409.1 |
- |
3 |
3033 |
|
- Gene Symbol
- Ebna1bp2
- Entrez Gene ID
- 69072
- Gene Name
- EBNA1 binding protein 2
- Gene Aliases
- 1810014B19Rik, AA537058, B830003A16Rik, C81321, Ebp2, Nobp, p40
- Chromosome Location
- Chr. 4: 118620799 - 118627776 on Build GRCm38
- UniGene ID
- Mm.29906
- Species
- Mus musculus
- Gene Symbol
- Esr1
- Entrez Gene ID
- 13982
- Gene Name
- estrogen receptor 1 (alpha)
- Gene Aliases
- ER, ER-alpha, ERa, ERalpha, ESR, Estr, Estra, Nr3a1
- Chromosome Location
- Chr. 10: 4611989 - 5005633 on Build GRCm38
- UniGene ID
- Mm.463262
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039911.1 |
- |
3 |
406 |
|
- Gene Symbol
- Fam169b
- Entrez Gene ID
- 434197
- Gene Name
- family with sequence similarity 169, member B
- Gene Aliases
- EG434197
- Chromosome Location
- Chr. 7: 68273839 - 68363089 on Build GRCm38
- UniGene ID
- Mm.216193
- Species
- Mus musculus
- Gene Symbol
- Fbxl17
- Entrez Gene ID
- 50758
- Gene Name
- F-box and leucine-rich repeat protein 17
- Gene Aliases
- 6330576B01Rik, AI452053, BB073797, C130023C01Rik, Fbx13, Fbxo13
- Chromosome Location
- Chr. 17: 63045949 - 63504042 on Build GRCm38
- UniGene ID
- Mm.136366
- Species
- Mus musculus
- Gene Symbol
- G630055G22Rik
- Entrez Gene ID
- 414127
- Gene Name
- RIKEN cDNA G630055G22 gene
- Gene Aliases
- Chromosome Location
- Chr. 18: 47922841 - 48107980 on Build GRCm38
- UniGene ID
- Mm.450768
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK148393.1 |
- |
1 |
32 |
|
- Gene Symbol
- Glrx2
- Entrez Gene ID
- 69367
- Gene Name
- glutaredoxin 2 (thioltransferase)
- Gene Aliases
- 1700010P22Rik, AI645710, Grx2
- Chromosome Location
- Chr. 1: 143739224 - 143749678 on Build GRCm38
- UniGene ID
- Mm.272727
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029540.1 |
- |
1 |
1004 |
|
- Gene Symbol
- Gm10002
- Entrez Gene ID
- 791405
- Gene Name
- predicted gene 10002
- Gene Aliases
- ENSMUSG00000056640
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.483576
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040372.1 |
- |
1 |
3050 |
|
- Gene Symbol
- Gm10433
- Entrez Gene ID
- 100038378
- Gene Name
- predicted gene 10433
- Gene Aliases
- ENSMUSG00000072884
- Chromosome Location
- Chr. 12: 100187968 - 100193538 on Build GRCm38
- UniGene ID
- Mm.387075
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141725.1 |
- |
3 |
1864 |
|
- Gene Symbol
- Gm10465
- Entrez Gene ID
- 100038648
- Gene Name
- predicted gene 10465
- Gene Aliases
- ENSMUSG00000073092
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.482648
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK142489.1 |
- |
1 |
32 |
|
- Gene Symbol
- Gm10522
- Entrez Gene ID
- 100038407
- Gene Name
- predicted gene 10522
- Gene Aliases
- ENSMUSG00000073493
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK154426.1 |
- |
1 |
32 |
|
- Gene Symbol
- Gm14204
- Entrez Gene ID
- 100045469
- Gene Name
- predicted gene 14204
- Gene Aliases
- OTTMUSG00000015935
- Chromosome Location
- Chr. 2: 158595472 - 158610723 on Build GRCm38
- UniGene ID
- Mm.143404
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK134637.1 |
- |
2 |
300 |
|
AK143055.1 |
- |
2 |
365 |
|
- Gene Symbol
- Gm14340
- Entrez Gene ID
- 676369
- Gene Name
- predicted gene 14340
- Gene Aliases
- OTTMUSG00000016394
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.388439
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK133152.1 |
- |
2 |
1596 |
|
- Gene Symbol
- Gm33395
- Entrez Gene ID
- 102636287
- Gene Name
- predicted gene, 33395
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK132357.1 |
- |
1 |
2478 |
|
- Gene Symbol
- Gm3367
- Entrez Gene ID
- 100041489
- Gene Name
- predicted gene 3367
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.383855
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AA822171.1 |
- |
|
24 |
|
- Gene Symbol
- Gm35387
- Entrez Gene ID
- 102638952
- Gene Name
- predicted gene, 35387
- Gene Aliases
- Chromosome Location
- Chr. 14: 29903768 - 29907588 on Build GRCm38
- UniGene ID
- Mm.298706
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC066010.1 |
- |
2 |
1701 |
|
- Gene Symbol
- Gm36447
- Entrez Gene ID
- 102640374
- Gene Name
- predicted gene, 36447
- Gene Aliases
- Chromosome Location
- Chr. 5: 150821881 - 150866601 on Build GRCm38
- UniGene ID
- Mm.366871
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136043.1 |
- |
4 |
1893 |
|
- Gene Symbol
- Gm38414
- Entrez Gene ID
- 330738
- Gene Name
- predicted gene, 38414
- Gene Aliases
- A230084N10
- Chromosome Location
- Chr. 8: 36156010 - 36161582 on Build GRCm38
- UniGene ID
- Mm.128075
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039009.1 |
- |
3 |
1284 |
|
- Gene Symbol
- Gm40170
- Entrez Gene ID
- 105244577
- Gene Name
- predicted gene, 40170
- Gene Aliases
- Chromosome Location
- Chr. 3: 146763280 - 146774114 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039285.1 |
- |
|
52 |
|
- Gene Symbol
- Gm41496
- Entrez Gene ID
- 105246162
- Gene Name
- predicted gene, 41496
- Gene Aliases
- Chromosome Location
- Chr. 16: 92125734 - 92189722 on Build GRCm38
- UniGene ID
- Mm.488330
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK087806.1 |
- |
3 |
1316 |
|
- Gene Symbol
- Gm6182
- Entrez Gene ID
- 100504263
- Gene Name
- predicted gene 6182
- Gene Aliases
- 2210418O10Rik
- Chromosome Location
- Chr. 2: 176226445 - 176237913 on Build GRCm38
- UniGene ID
- Mm.379980
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC080301.1 |
- |
|
4733 |
|
BC090643.1 |
- |
|
4465 |
|
- Gene Symbol
- Gm7969
- Entrez Gene ID
- 666185
- Gene Name
- predicted gene 7969
- Gene Aliases
- Chromosome Location
- Chr. 13: 67949751 - 67954507 on Build GRCm38
- UniGene ID
- Mm.425830
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK139503.1 |
- |
3 |
2409 |
|
- Gene Symbol
- Gm9924
- Entrez Gene ID
- 791336
- Gene Name
- predicted gene 9924
- Gene Aliases
- ENSMUSG00000053839
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.404552
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042950.1 |
- |
1 |
2193 |
|
- Gene Symbol
- LOC105245566
- Entrez Gene ID
- 105245566
- Gene Name
- uncharacterized LOC105245566
- Gene Aliases
- Chromosome Location
- Chr. 13: 81782594 - 81783769 on Build GRCm38
- UniGene ID
- Mm.401912
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK161157.1 |
- |
|
2917 |
|
- Gene Symbol
- LOC544724
- Entrez Gene ID
- 544724
- Gene Name
- uncharacterized LOC544724
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.393362
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK085787.1 |
- |
1 |
330 |
|
- Gene Symbol
- Lars2
- Entrez Gene ID
- 102436
- Gene Name
- leucyl-tRNA synthetase, mitochondrial
- Gene Aliases
- AI035546, Kiaa0028, LEURS
- Chromosome Location
- Chr. 9: 123366940 - 123462666 on Build GRCm38
- UniGene ID
- Mm.276076
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK035538.1 |
- |
2 |
1996 |
|
- Gene Symbol
- Mmp16
- Entrez Gene ID
- 17389
- Gene Name
- matrix metallopeptidase 16
- Gene Aliases
- MT3-MMP, Mt3mmp
- Chromosome Location
- Chr. 4: 17853482 - 18118734 on Build GRCm38
- UniGene ID
- Mm.187315
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077939.1 |
- |
|
1548 |
|
- Gene Symbol
- Nek4
- Entrez Gene ID
- 23955
- Gene Name
- NIMA (never in mitosis gene a)-related expressed kinase 4
- Gene Aliases
- Stk2
- Chromosome Location
- Chr. 14: 30951382 - 30988476 on Build GRCm38
- UniGene ID
- Mm.251494
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK149147.1 |
- |
6 |
2115 |
|
AK149584.1 |
- |
14 |
3831 |
|
- Gene Symbol
- Neurl3
- Entrez Gene ID
- 214854
- Gene Name
- neuralized homolog 3 homolog (Drosophila)
- Gene Aliases
- 2010300P06Rik, Lincr
- Chromosome Location
- Chr. 1: 36264602 - 36274674 on Build GRCm38
- UniGene ID
- Mm.389110
- Species
- Mus musculus
- Gene Symbol
- Nipal3
- Entrez Gene ID
- 74552
- Gene Name
- NIPA-like domain containing 3
- Gene Aliases
- 9130020G22Rik, AI480660, Npal3
- Chromosome Location
- Chr. 4: 135447136 - 135494806 on Build GRCm38
- UniGene ID
- Mm.26548
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK053937.1 |
- |
|
2513 |
|
- Gene Symbol
- Plcl2
- Entrez Gene ID
- 224860
- Gene Name
- phospholipase C-like 2
- Gene Aliases
- PLC-L2, PRIP-2, Plce2, mKIAA1092
- Chromosome Location
- Chr. 17: 50508633 - 50688494 on Build GRCm38
- UniGene ID
- Mm.217362
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK163182.1 |
- |
|
3615 |
|
- Gene Symbol
- Smlr1
- Entrez Gene ID
- 100504474
- Gene Name
- small leucine-rich protein 1
- Gene Aliases
- 2010003K15Rik
- Chromosome Location
- Chr. 10: 25527943 - 25536272 on Build GRCm38
- UniGene ID
- Mm.46323
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK080199.1 |
- |
1 |
3224 |
|
- Gene Symbol
- Tprkb
- Entrez Gene ID
- 69786
- Gene Name
- Tp53rk binding protein
- Gene Aliases
- 0610033G21Rik, 1810034M08Rik, Cgi-121, DRWMS1, mDRWMS1
- Chromosome Location
- Chr. 6: 85915278 - 85930284 on Build GRCm38
- UniGene ID
- Mm.218434
- Species
- Mus musculus
- Gene Symbol
- Trim43a
- Entrez Gene ID
- 547109
- Gene Name
- tripartite motif-containing 43A
- Gene Aliases
- EG547109, Gm6021
- Chromosome Location
- Chr. 9: 88580750 - 88588823 on Build GRCm38
- UniGene ID
- Mm.304302
- Species
- Mus musculus
- Gene Symbol
- Trim43b
- Entrez Gene ID
- 666747
- Gene Name
- tripartite motif-containing 43B
- Gene Aliases
- EG666747, Gm8269
- Chromosome Location
- Chr. 9: 89084624 - 89092835 on Build GRCm38
- UniGene ID
- Mm.425592
- Species
- Mus musculus
- Gene Symbol
- Trim43c
- Entrez Gene ID
- 666731
- Gene Name
- tripartite motif-containing 43C
- Gene Aliases
- EG666731, Trim43
- Chromosome Location
- Chr. 9: 88839023 - 88848198 on Build GRCm38
- UniGene ID
- Mm.446951
- Species
- Mus musculus
- Gene Symbol
- Zfp444
- Entrez Gene ID
- 72667
- Gene Name
- zinc finger protein 444
- Gene Aliases
- 2810031J10Rik, 6230401O10Rik, mFLJ00134
- Chromosome Location
- Chr. 7: 6172462 - 6193104 on Build GRCm38
- UniGene ID
- Mm.274089
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK220307.1 |
- |
|
4237 |
|
- Gene Symbol
- Zfp862-ps
- Entrez Gene ID
- 58894
- Gene Name
- zinc finger protein 862, pseudogene
- Gene Aliases
- 4732460K03Rik, AI448302, Zfp862, mKIAA0543
- Chromosome Location
- Chr. 6: 48504339 - 48534832 on Build GRCm38
- UniGene ID
- Mm.31983
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK166322.1 |
- |
4 |
1821 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK038577.1 |
- |
|
1131 |
|
Chr. - - [-] |
GenBankmRNA |
AK037041.1 |
- |
|
32 |
|
Chr. - - [-] |
GenBankmRNA |
AK164664.1 |
- |
|
1739 |
|
Chr. - - [-] |
GenBankmRNA |
AK086884.1 |
- |
|
2909 |
|
Chr. - - [-] |
GenBankmRNA |
AK031173.1 |
- |
|
33 |
|
Chr. - - [-] |
GenBankmRNA |
AK136139.1 |
- |
|
32 |
|
Chr. - - [-] |
View Full Product Details