Gene Transcripts

Gene Symbol
Nup214
Entrez Gene ID
227720
Gene Name
nucleoporin 214
Gene Aliases
BC039282, CAN, D2H9S46E
Chromosome Location
Chr. 2: 31974420 - 32053975 on Build GRCm38
UniGene ID
Mm.289686
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172268.2 NP_758472.2 22 3019
GenBank mRNA AK122189.1 - 22 2988
BC039282.1 - 12 1376 4976266

View Full Product Details