Gene Transcripts
- Gene Symbol
- Naa35
- Entrez Gene ID
- 78689
- Gene Name
- N(alpha)-acetyltransferase 35, NatC auxiliary subunit
- Gene Aliases
- A330021G12Rik, A330027C19Rik, AI158944, C030004C14Rik, Mak10
- Chromosome Location
- Chr. 13: 59585323 - 59634798 on Build GRCm38
- UniGene ID
- Mm.253902
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039339.1 |
- |
1 |
660 |
|
AK052852.1 |
- |
1 |
596 |
|
View Full Product Details