Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000144.4 | NP_000135.2 | 4 | 678 | |
NM_001161706.1 | NP_001155178.1 | 4 | 678 | ||
NM_181425.2 | NP_852090.1 | 4 | 678 | ||
GenBank mRNA | AK308620.1 | - | 4 | 338 | |
BC023633.2 | - | 4 | 486 | 4842134 | |
BC048097.1 | - | 4 | 489 | 5300379 | |
BF058880.1 | - | 4 | 349 | ||
KU178079.1 | - | 4 | 458 | ||
KU178080.1 | - | 3 | 337 | ||
U43747.1 | - | 4 | 983 |