Gene Transcripts

Gene Symbol
FBXO44
Entrez Gene ID
93611
Gene Name
F-box protein 44
Gene Aliases
FBG3, FBX30, FBX6A, Fbx44, Fbxo6a
Chromosome Location
Chr. 1: 11654375 - 11663327 on Build GRCh38
UniGene ID
Hs.556006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014765.1 NP_001014765.1 7 1229
NM_001304790.1 NP_001291719.1 6 762
NM_001304791.1 NP_001291720.1 6 968
NM_033182.5 NP_149438.2 6 857
NM_183412.2 NP_904319.1 5 843
NM_183413.2 NP_904320.1 5 732
NR_130909.1 5 598
XM_005263535.1 XP_005263592.1 6 799
XM_005263536.4 XP_005263593.1 7 1193
XM_005263537.1 XP_005263594.1 6 939
XM_006711043.2 XP_006711106.1 6 799
XM_006711045.2 XP_006711108.1 6 830
XM_011542435.1 XP_011540737.1 7 1261
XM_017002842.1 XP_016858331.1 6 799
XM_017002843.1 XP_016858332.1 6 830
XM_017002844.1 XP_016858333.1 5 703
GenBank mRNA AK055344.1 - 5 843
AK293721.1 - 6 776
AK296025.1 - 5 578
AK301418.1 - 6 837
AK316284.1 - 5 976
AK316347.1 - 6 814
AY007380.1 - 4 639
AY040878.1 - 6 813
BC007832.2 - 5 679 4054414
BU164151.1 - 6 686
CR749368.1 - 7 1218

View Full Product Details