Gene Transcripts

Gene Symbol
LINC00661
Entrez Gene ID
126536
Gene Name
long intergenic non-protein coding RNA 661
Gene Aliases
Chromosome Location
Chr. 19: 16015634 - 16027462 on Build GRCh38
UniGene ID
Hs.131824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026828.1 7 715
GenBank mRNA AK128240.1 - 2 213
BC117476.1 - 3 317
BC144048.1 - 3 317
CR749400.1 - 7 715

View Full Product Details