Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199427.1 | NP_001186356.1 | 7 | 955 | |
NM_003300.3 | NP_003291.2 | 10 | 1204 | ||
NM_145725.2 | NP_663777.1 | 11 | 1343 | ||
NM_145726.2 | NP_663778.1 | 10 | 1268 | ||
XM_011537116.2 | XP_011535418.1 | 9 | 1163 | ||
XM_011537117.2 | XP_011535419.1 | 9 | 1151 | ||
XM_011537118.2 | XP_011535420.1 | 8 | 1071 | ||
XM_017021617.1 | XP_016877106.1 | 11 | 1763 | ||
XM_017021618.1 | XP_016877107.1 | 11 | 3259 | ||
XM_017021619.1 | XP_016877108.1 | 10 | 3186 | ||
GenBank mRNA | AK303172.1 | - | 7 | 832 | |
BC075086.2 | - | 9 | 1015 | 30915387 | |
BC075087.2 | - | 9 | 1015 | 30915599 | |
BX247977.1 | - | 11 | 1189 | ||
L38509.1 | - | 9 | 1051 | ||
U15637.1 | - | 11 | 1197 | ||
U19260.1 | - | 10 | 1140 | ||
U21092.1 | - | 10 | 1207 |