Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003841.3 | NP_003832.2 | 3 | 529 | |
GenBank mRNA | AF012536.1 | - | 3 | 413 | |
AF012629.1 | - | 3 | 221 | ||
AF014794.1 | - | 3 | 370 | ||
AF016267.1 | - | 3 | 408 | ||
AF020502.1 | - | 3 | 341 | ||
AF033854.1 | - | 3 | 397 | ||
AK298664.1 | - | 2 | 316 | ||
AK298718.1 | - | 3 | 420 | ||
AK313213.1 | - | 3 | 423 | ||
AY358281.1 | - | 3 | 413 | ||
BC017852.1 | - | 3 | 422 | ||
BC125041.1 | - | 3 | 342 | 40115748 | |
BC125042.1 | - | 3 | 342 | 40115749 | |
CR541857.1 | - | 3 | 221 | ||
DC380668.1 | - | 3 | 529 |