Gene Transcripts

Gene Symbol
Robo1
Entrez Gene ID
19876
Gene Name
roundabout homolog 1 (Drosophila)
Gene Aliases
AW494633, AW742721, DUTT1, Gm310
Chromosome Location
Chr. 16: 72027553 - 73047491 on Build GRCm38
UniGene ID
Mm.310772
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019413.2 NP_062286.2 14 3032
GenBank mRNA AK077498.1 - 2 1529
Y17793.1 - 14 2294

View Full Product Details