Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168509.1 | NP_001161981.1 | 3 | 793 | |
NM_001168510.1 | NP_001161982.1 | 3 | 730 | ||
NM_001168511.1 | NP_001161983.1 | 3 | 737 | ||
NM_001168512.1 | NP_001161984.1 | 2 | 883 | ||
NM_146187.4 | NP_666299.1 | 3 | 1116 | ||
GenBank mRNA | AF545043.1 | - | 3 | 667 | |
AK050524.1 | - | 1 | 748 | ||
AK078861.1 | - | 3 | 1116 | ||
BC019570.1 | - | 1 | 793 | 4218874 |