Gene Transcripts

Gene Symbol
Zmiz1
Entrez Gene ID
328365
Gene Name
zinc finger, MIZ-type containing 1
Gene Aliases
BC065120, E330020C23, ENSMUSG00000072684, Gm10397, I920194n01, Rai17, Zimp10
Chromosome Location
Chr. 14: 25458865 - 25666747 on Build GRCm38
UniGene ID
Mm.227484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183208.3 NP_899031.2 21 3113
GenBank mRNA BC057691.1 - 11 1350
BC058646.1 - 21 3013 6405777
BC065120.1 - 21 3006 6856060

View Full Product Details