Gene Transcripts

Gene Symbol
Armc3
Entrez Gene ID
100361506
Gene Name
armadillo repeat containing 3
Gene Aliases
Chromosome Location
Chr. 17: 85914587 - 86009403 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287020.1 NP_001273949.1 6 536

View Full Product Details