Gene Transcripts

Gene Symbol
Zmiz1
Entrez Gene ID
361103
Gene Name
zinc finger, MIZ-type containing 1
Gene Aliases
Rai17
Chromosome Location
Chr. 16: 1749126 - 1954633 on Build Rnor_6.0
UniGene ID
Rn.1712
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108393.1 NP_001101863.1 23 3606
XM_006252537.2 XP_006252599.1 22 16117
XM_006252538.2 XP_006252600.1 23 3653

View Full Product Details