Gene Transcripts

Gene Symbol
Ankle1
Entrez Gene ID
361122
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
Ankrd41, RGD1308184
Chromosome Location
Chr. 16: 19800390 - 19826057 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252963.2 XP_006253025.1 2 844

View Full Product Details