Gene Transcripts

Gene Symbol
Gas6
Entrez Gene ID
58935
Gene Name
growth arrest specific 6
Gene Aliases
Chromosome Location
Chr. 16: 81213364 - 81243757 on Build Rnor_6.0
UniGene ID
Rn.52228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057100.2 NP_476441.2 5, 6 580
GenBank mRNA BC070881.1 - 5, 6 580
D42148.1 - 5, 6 475

View Full Product Details