Gene Transcripts

Gene Symbol
Tsx
Entrez Gene ID
29391
Gene Name
testis specific X-linked gene
Gene Aliases
Chromosome Location
Chr. X: 74269513 - 74283819 on Build Rnor_6.0
UniGene ID
Rn.87406
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019203.1 NP_062076.1 5 386
XM_006257142.2 XP_006257204.1 6 466
GenBank mRNA X99797.1 - 5 386

View Full Product Details