Gene Transcripts
- Gene Symbol
- LOC100128288
- Entrez Gene ID
- 100128288
- Gene Name
- uncharacterized LOC100128288
- Gene Aliases
- Chromosome Location
- Chr. 17: 8358413 - 8360541 on Build GRCh38
- UniGene ID
- Hs.549913
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024447.1 |
|
1 |
1371 |
|
GenBank mRNA |
AK094860.1 |
- |
1 |
1050 |
|
View Full Product Details