Gene Transcripts
- Gene Symbol
- LINC01152
- Entrez Gene ID
- 102606463
- Gene Name
- long intergenic non-protein coding RNA 1152
- Gene Aliases
- CMPD, TCONS_00025128
- Chromosome Location
- Chr. 17: 72030654 - 72041297 on Build GRCh38
- UniGene ID
- Hs.512309
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110124.1 |
|
|
3 |
602 |
|
GenBank mRNA |
D43770.1 |
- |
LIT1584 CMPD |
3 |
440 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000472655.3 |
- |
|
602 |
|
Chr. 17 - 72030654 - 72038265 [+] |
View Full Product Details