Gene Transcripts
- Gene Symbol
- USP33
- Entrez Gene ID
- 23032
- Gene Name
- ubiquitin specific peptidase 33
- Gene Aliases
- VDU1
- Chromosome Location
- Chr. 1: 77695987 - 77759879 on Build GRCh38
- UniGene ID
- Hs.480597
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094056.1 |
- |
1 |
288 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB257060.1 |
- |
1 |
289 |
|
Chr. 1 - 77697360 - 77697937 [-] |
GenBankmRNA |
DA362717.1 |
- |
2 |
450 |
|
Chr. 1 - 77697547 - 77701536 [-] |
View Full Product Details