Gene Transcripts
- Gene Symbol
- 2810019C22Rik
- Entrez Gene ID
- 72633
- Gene Name
- RIKEN cDNA 2810019C22 gene
- Gene Aliases
- AI451161
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.32874
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK012763.1 |
- |
1 |
39 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY711879.1 |
- |
1 |
37 |
|
Chr. 9 - 88448043 - 88448947 [-] |
View Full Product Details