Gene Transcripts
- Gene Symbol
- Gm16499
- Entrez Gene ID
- 100049166
- Gene Name
- predicted gene 16499
- Gene Aliases
- ENSMUSG00000068503
- Chromosome Location
- Chr. 6: 53844899 - 53867140 on Build GRCm38
- UniGene ID
- Mm.404560
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK039090.1 |
- |
|
2024 |
|
View Full Product Details