Gene Transcripts
- Gene Symbol
- Mro
- Entrez Gene ID
- 71263
- Gene Name
- maestro
- Gene Aliases
- 4930507C04Rik, 4933435E20Rik
- Chromosome Location
- Chr. 18: 73851827 - 73881334 on Build GRCm38
- UniGene ID
- Mm.126220
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015725.1 |
- |
4 |
611 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY715266.1 |
- |
4 |
611 |
|
Chr. 18 - 73859394 - 73881332 [+] |
View Full Product Details